Learn More
Abnova™ Human FBXO6 Full-length ORF (AAH20880.1, 1 a.a. - 293 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | AAH20880.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26270 |
Molecular Weight (g/mol) | 57.97kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-944-257
|
Abnova™
H00026270P01S |
10 ug |
Each for $499.00
|
|
|||||
89-944-258
|
Abnova™
H00026270P01L |
25 ug |
Each for $769.00
|
|
|||||
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class, and its C-terminal region is highly similar to that of rat NFB42 (neural F Box 42 kDa) which may be involved in the control of the cell cycle. [provided by RefSeq]
Sequence: MDAPHSKAALDSINELPENILLELFTHVPARQLLLNCRLVCSLWRDLIDLMTLWKRKCLREGFITKDWDQPVADWKIFYFLRSLHRNLLRNPCAEEDMFAWQIDFNGGDRWKVESLPGAHGTDFPDPKVKKYFVTSYEMCLKSQLVDLVAEGYWEELLDTFRPDIVVKDWFAARADCGCTYQLKVQLASADYFVLASFEPPPVTIQQWNNATWTEVSYTFSDYPRGVRYILFQHGGRDTQYWAGWYGPRVTNSSIVVSPKMTRNQASSEAQPGQKHGQEEAAQSPYRAVVQIFSpecifications
AAH20880.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.97kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDAPHSKAALDSINELPENILLELFTHVPARQLLLNCRLVCSLWRDLIDLMTLWKRKCLREGFITKDWDQPVADWKIFYFLRSLHRNLLRNPCAEEDMFAWQIDFNGGDRWKVESLPGAHGTDFPDPKVKKYFVTSYEMCLKSQLVDLVAEGYWEELLDTFRPDIVVKDWFAARADCGCTYQLKVQLASADYFVLASFEPPPVTIQQWNNATWTEVSYTFSDYPRGVRYILFQHGGRDTQYWAGWYGPRVTNSSIVVSPKMTRNQASSEAQPGQKHGQEEAAQSPYRAVVQIF | |
RUO | |
FBXO6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
26270 | |
FBXO6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBG2/FBS2/FBX6/Fbx6b | |
FBXO6 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.