Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FBXO8 Full-length ORF (NP_036312.2, 1 a.a. - 319 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name FBXO8 (Human) Recombinant Protein (P02)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 63.5kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-944-268
View Documents
Abnova™
H00026269P02L
25 ug
Each for $709.50
Add to cart
 
89-944-267
View Documents
Abnova™
H00026269P02S
10 ug
Each for $460.50
Add to cart
 
Description

Description

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It contains a C-terminal amino acid sequence that bears a significant similarity with a portion of yeast Sec7p, a critical regulator of vesicular protein transport. This human protein may interact with ADP-ribosylation factor(s)(ARFs) and exhibit ARF-GEF (guanine nucleotide exchange factor) activity. [provided by RefSeq]

Sequence: MGQGLWRVVRNQQLQQEGYSEQGYLTREQSRRMAASNISNTNHRKQVQGGIDIYHLLKARKSKEQEGFINLEMLPPELSFTILSYLNATDLCLASCVWQDLANDELLWQGLCKSTWGHCSIYNKNPPLGFSFRKLYMQLDEGSLTFNANPDEGVNYFMSKGILDDSPKEIAKFIFCTRTLNWKKLRIYLDERRDVLDDLVTLHNFRNQFLPNALREFFRHIHAPEERGEYLETLITKFSHRFCACNPDLMRELGLSPDAVYVLCYSLILLSIDLTSPHVKNKMSKREFIRNTRRAAQNISEDFVGHLYDNIYLIGHVAA
Specifications

Specifications

FBXO8 (Human) Recombinant Protein (P02)
Glutathione Sepharose 4 Fast Flow
63.5kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit