Learn More
Abnova™ Human FBXW8 Partial ORF (NP_699179, 499 a.a. - 598 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00026259Q01L
Description
This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene contains a WD-40 domain, in addition to an F-box motif, so it belongs to the Fbw class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: VWDYRMNQKLWEVYSGHPVQHISFSSHSLITANVPYQTVMRNADLDSFTTHRRHRGLIRAYEFAVDQLAFQSPLPVCRSSCDAMATHYYDLALAFPYNHVSpecifications
NP_699179 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VWDYRMNQKLWEVYSGHPVQHISFSSHSLITANVPYQTVMRNADLDSFTTHRRHRGLIRAYEFAVDQLAFQSPLPVCRSSCDAMATHYYDLALAFPYNHV | |
RUO | |
FBXW8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
26259 | |
FBXW8 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBW6/FBW8/FBX29/FBXO29/FBXW6/MGC33534 | |
FBXW8 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.