Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fc gamma RIIIA (CD16a) (176 Phe) His-tag, Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

HEK293 cells Recombinant Protein

$534.00

Specifications

Recombinant Yes
Name SARS-CoV-2 Spike Protein (S-RBD) (aa330-524), His Tag
For Use With (Application) Control, ELISA, Western Blot
Conjugate Unlabeled
Format Lyophilized
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87685 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87685
50 µg
Each for $534.00
Add to cart
 
Description

Description

CD16 (FCGR3A) is a 50-65 kDa cell surface molecule that exists in two forms - a transmembranous form expressed by NK cells and some T cells, and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC by NK cells. Human CD16 is expressed in two forms – FCGR3A and FCGR3B. FCGR3A is associated with the FcepsilonRI-gamma subunit and is responsible for antibody-dependent NK cell cytotoxicity. Mast cell FCGR3A is associated, with FcepsilonRI-beta subunit. Besides IgG, FCGR3A can be triggered also by oligomeric IgE. FCGR3B is a GPI-linked monomeric receptor expressed on neutrophils and is involved in their activation and induction of a pro-adhesive phenotype. Diseases associated with CD16 dysfunction include immunodeficiency 20 and systemic lupus erythematosus.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Lyophilized
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
RUO
Virus
SARS-CoV-2 Spike Protein (S-RBD) (aa330-524), His Tag
Unlabeled
PBS with no preservative
HEK293 cells
-20°Cor to +80°C if preferred
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit