Learn More
Abnova™ Human FCAMR Full-length ORF (NP_114418.2) Recombinant Protein without tag
Used for AP, Func, Screening
Supplier: Abnova™ H00083953G01
Description
This gene encodes a member of the immunoglobulin receptor gene family involved in the microbial immune response. The protein localizes to the cell surface, where it functions as a receptor for IgM and IgA antibodies. The mouse homolog of this protein can bind and internalize IgM-coated particles, and mediates endocytosis of IgM-coated Staphylococcus by primary B lymphocytes. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Sequence: MDGEATVKPGEQKEVVRRGREVDYSRLIAGTLPQSHVTSRRAGWKMPLFLILCLLQGSSFALPQKRPHPRWLWEGSLPSRTHLRAMGTLRPSSPLCWREESSFAAPNSLKGSRLVSGEPGGAVTIQCHYAPSSVNRHQRKYWCRLGPPRWICQTIVSTNQYTHHRYRDRVALTDFPQRGLFVVRLSQLSPDDIGCYLCGIGSENNMLFLSMNLTISAVLFQKMKAALGPWLLSLPCWPCLCLWLWFYCKGSSGEGGPLRRQKGSPSpecifications
NP_114418.2 | |
Liquid | |
29.15kDa | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FCA/MR/FKSG87 | |
FCAMR | |
Recombinant | |
wheat germ expression system with proprietary liposome technology |
Antibody Production, Compound Screening, Functional Study | |
83953 | |
FCAMR (Human) Recombinant Protein | |
MDGEATVKPGEQKEVVRRGREVDYSRLIAGTLPQSHVTSRRAGWKMPLFLILCLLQGSSFALPQKRPHPRWLWEGSLPSRTHLRAMGTLRPSSPLCWREESSFAAPNSLKGSRLVSGEPGGAVTIQCHYAPSSVNRHQRKYWCRLGPPRWICQTIVSTNQYTHHRYRDRVALTDFPQRGLFVVRLSQLSPDDIGCYLCGIGSENNMLFLSMNLTISAVLFQKMKAALGPWLLSLPCWPCLCLWLWFYCKGSSGEGGPLRRQKGSP | |
RUO | |
FCAMR | |
Wheat Germ (in vitro) | |
None | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.