Learn More
Abnova™ Human FCGR2A (P12318, Gln 34 - Gly 216) Partial Recombinant Protein with C-terminal His tag expressed in HEK293 cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P3684
Catalog No. 89-944-310
Add to cart
Description
Description
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants. [provided by RefSeq]