Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Abnova™ Human FCGR2B Full-length ORF (NP_003992.3) Recombinant Protein without tag
Used for AP, Func, Screening
Manufacturer:  Abnova™ H00002213G01
Catalog No. 89-944-318
Description
Description
Sequence: MGILSFLPVLATESDWADCKSPQPWGHMLLWTAVLFLAPVAGTPAAPPKAVLKLEPQWINVLQEDSVTLTCRGTHSPESDSIQWFHNGNLIPTHTQPSYRFKANNNDSGEYTCQTGQTSLSDPVHLTVLSEWLVLQTPHLEFQEGETIVLRCHSWKDKPLVKVTFFQNGKSKKFSRSDPNFSIPQANHSHSGDYHCTGNIGYTLYSSKPVTITVQAPSSSPMGIIVAVVTGIAVAAIVAAVVALIYCRKKRISALPGYPECREMGETLPEKPANPTNPDEADKVGAENTITYSLLMHPDALEEPDDQNRI