Learn More
Abnova™ Human FCN1 Full-length ORF (NP_001994.2, 1 a.a. - 326 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00002219P01S
Description
The ficolin family of proteins are characterized by the presence of a leader peptide, a short N-terminal segment, followed by a collagen-like region, and a C-terminal fibrinogen-like domain. The collagen-like and the fibrinogen-like domains are also found separately in other proteins such as complement protein C1q, C-type lectins known as collectins, and tenascins. However, all these proteins recognize different targets, and are functionally distinct. Ficolin 1 encoded by FCN1 is predominantly expressed in the peripheral blood leukocytes, and has been postulated to function as a plasma protein with elastin-binding activity. [provided by RefSeq]
Sequence: MELSGATMARGLAVLLVLFLHIKNLPAQAADTCPEVKVVGLEGSDKLTILRGCPGLPGAPGPKGEAGVIGERGERGLPGAPGKAGPVGPKGDRGEKGMRGEKGDAGQSQSCATGPRNCKDLLDRGYFLSGWHTIYLPDCRPLTVLCDMDTDGGGWTVFQRRMDGSVDFYRDWAAYKQGFGSQLGEFWLGNDNIHALTAQGSSELRVDLVDFEGNHQFAKYKSFKVADEAEKYKLVLGAFVGGSAGNSLTGHNNNFFSTKDQDNDVSSSNCAEKFQGAWWYADCHASNLNGLYLMGPHESYANGINWSAAKGYKYSYKVSEMKVRPASpecifications
NP_001994.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FCNM | |
FCN1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2219 | |
FCN1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MELSGATMARGLAVLLVLFLHIKNLPAQAADTCPEVKVVGLEGSDKLTILRGCPGLPGAPGPKGEAGVIGERGERGLPGAPGKAGPVGPKGDRGEKGMRGEKGDAGQSQSCATGPRNCKDLLDRGYFLSGWHTIYLPDCRPLTVLCDMDTDGGGWTVFQRRMDGSVDFYRDWAAYKQGFGSQLGEFWLGNDNIHALTAQGSSELRVDLVDFEGNHQFAKYKSFKVADEAEKYKLVLGAFVGGSAGNSLTGHNNNFFSTKDQDNDVSSSNCAEKFQGAWWYADCHASNLNGLYLMGPHESYANGINWSAAKGYKYSYKVSEMKVRPA | |
RUO | |
FCN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.