Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FCRL3 Partial ORF (NP_001019838, 625 a.a. - 723 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00115352Q01S

 View more versions of this product

Catalog No. 89-944-344


Add to cart

Description

Description

This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains immunoreceptor-tyrosine activation motifs and immunoreceptor-tyrosine inhibitory motifs in its cytoplasmic domain and may play a role in regulation of the immune system. Mutations in this gene have been associated with rheumatoid arthritis, autoimmune thyroid disease, and systemic lupus erythematosus. [provided by RefSeq]

Sequence: SRPSRIDPQEPTHSKPLAPMELEPMYSNVNPGDSNPIYSQIWSIQHTKENSANCPMMHQEHEELTVLYSELKKTHPDDSAGEASSRGRAHEEDDEENYE
Specifications

Specifications

NP_001019838
Liquid
115352
FCRL3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FCRH3|IFGP3|IRTA3|SPAP2
FCRL3
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SRPSRIDPQEPTHSKPLAPMELEPMYSNVNPGDSNPIYSQIWSIQHTKENSANCPMMHQEHEELTVLYSELKKTHPDDSAGEASSRGRAHEEDDEENYE
RUO
FCRL3
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit