Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FCRLB Full-length ORF (AAH67080.1, 1 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00127943P01

Catalog No. 89-944-352


Add to cart

Description

Description

FCRL2 belongs to the Fc receptor family. Fc receptors are involved in phagocytosis, antibody-dependent cell cytotoxicity, immediate hypersensitivity, and transcytosis of immunoglobulins via their ability to bind immunoglobulin (Ig) constant regions (Chikaev et al., 2005 [PubMed 15676285]).[supplied by OMIM]

Sequence: MSIEKFWDRSSARKERAEVQIAATWRVSLRLWTLSPLLCREAALDPPPAWRLVPFPDAHPGFSLWLGTPTERAALAGAGATVGLGRRSDSLASCKAGAASPILGCRTRAECQSGCDMKRLAWSLFLSSFPRPSWTRSPCPTFQKAPLPPPRGHTASSPPPLVCGSARSPRGKQEGAGSRSGGQAGAVAPRPLRRAVPGAGAEGDGSAGGPRRGAGWGGAALRHAPAPAEARHAAAVRVLQVQPRGAPLRLGRRVHSPGARGRGARIVLVRGGYRHPQCPETQSVAAAPGARFSPGPGLHHRPSSMGRSLGSW
Specifications

Specifications

AAH67080.1
Liquid
127943
FCRLB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSIEKFWDRSSARKERAEVQIAATWRVSLRLWTLSPLLCREAALDPPPAWRLVPFPDAHPGFSLWLGTPTERAALAGAGATVGLGRRSDSLASCKAGAASPILGCRTRAECQSGCDMKRLAWSLFLSSFPRPSWTRSPCPTFQKAPLPPPRGHTASSPPPLVCGSARSPRGKQEGAGSRSGGQAGAVAPRPLRRAVPGAGAEGDGSAGGPRRGAGWGGAALRHAPAPAEARHAAAVRVLQVQPRGAPLRLGRRVHSPGARGRGARIVLVRGGYRHPQCPETQSVAAAPGARFSPGPGLHHRPSSMGRSLGSW
RUO
FCRLB
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
59.1kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FCRL2|FCRLM2|FCRLY|FLJ31052|FREB-2|FcRY|RP11-474I16.6
FCRLB
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit