Learn More
Abnova™ Human FER (NP_005237.1, 1 a.a. - 822 a.a.) Full-lenth Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5711
Catalog No. 89-944-387
Add to cart
Description
Description
Fer protein is a member of the FPS/FES family of nontransmembrane receptor tyrosine kinases. It regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors. [provided by RefSeq]
Specifications
Specifications
NP_005237.1
Liquid
2241
FER (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TYK3
FER
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : Srctide. ATP: 100 µM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
122kDa
Baculovirus infected insect cell (Sf21) expression system
94% by SDS-PAGE/CBB staining.
5 ug
RUO
FER
Baculovirus infected insect cell (Sf21) expression system
Insect
GST