Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FER1L3 Partial ORF (NP_038479, 655 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026509Q01S

 View more versions of this product

Catalog No. 89-944-388


Add to cart

Description

Description

Mutations in dysferlin, a protein associated with the plasma membrane, can cause muscle weakness that affects both proximal and distal muscles. The protein encoded by this gene is a type II membrane protein that is structurally similar to dysferlin. It is a member of the ferlin family and associates with both plasma and nuclear membranes. The protein contains C2 domains that play a role in calcium-mediated membrane fusion events, suggesting that it may be involved in membrane regeneration and repair. Two transcript variants encoding different isoforms have been found for this gene. Other possible variants have been detected, but their full-length nature has not been determined. [provided by RefSeq]

Sequence: DAVNTLLAMAERLQTNIEALKSGIQGKIPANQLAELWLKLIDEVIEDTRYTLPLTEGKANVTVLDTQIRKLRSRSLSQIHEAAVRMRSEATDVKSTLAEI
Specifications

Specifications

NP_038479
Liquid
26509
FER1L3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FER1L3|FLJ36571|FLJ90777
MYOF
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DAVNTLLAMAERLQTNIEALKSGIQGKIPANQLAELWLKLIDEVIEDTRYTLPLTEGKANVTVLDTQIRKLRSRSLSQIHEAAVRMRSEATDVKSTLAEI
RUO
MYOF
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit