Learn More
Abnova™ Human FES (NM_002005) Full-length Recombinant Protein with GST-tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3861
Catalog No. 89-944-395
Add to cart
Description
Description
This gene encodes the human cellular counterpart of a feline sarcoma retrovirus protein with transforming capabilities. The gene product has tyrosine-specific protein kinase activity and that activity is required for maintenance of cellular transformation. Its chromosomal location has linked it to a specific translocation event identified in patients with acute promyelocytic leukemia but it is also involved in normal hematopoiesis as well as growth factor and cytokine receptor signaling. Alternative splicing results in multiple variants encoding different isoforms
Specifications
Specifications
NM_002005
In 50mM Tris-HCl, 150mM NaCl, pH 7.5 (0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol)
125kDa
Insect cell (Sf9) expression system
SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FPS
FES
The specific activity was determined to be 56nmol /min/mg as per Activity Assay Protocol.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
2242
FES (Human) Recombinant Protein
>90% by densitometry
5 ug
RUO
FES
Insect cell (Sf9) expression system
Insect
GST