Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FETUB Full-length ORF (AAH74734.1, 1 a.a. - 382 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026998P01S

 View more versions of this product

Catalog No. 89-944-397


Add to cart

Description

Description

The protein encoded by this gene is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. This protein may be secreted by cells. [provided by RefSeq]

Sequence: MGLLLPLAPCILVLCCGAMSPPQLALNPSALLSRGCNDSDVLAVAGFALRDINKDRKDGYVLRLNRVNDAQEYRRGGLGSLFYLTLDVLETDCHVLRKKAWQDCGMRIFFESVYGQCKAIFYMNNPSRVLYLAAYNCTLRPVSKKKIYMTCPDCPSSIPTDSSNHQVLEAATESLAKYKNENTSKQYSLFKVTRASSQWVVGPSYFVEYLIKESPCTKSQASSCSLQSSDSVPVGLCKGSLTRTHWEKFVSVTCDFFESQAPATGSENSAVNQKPTNLPKVEESQQKNTPPTDSPSKAGPRGSVQYLPDLDDKNSQEKGPQEAFPVHLDLTTNPQGETLDISFLFLEPMEEKLVVLPFPKEKARTAECPGPAQNASPLVLPP
Specifications

Specifications

AAH74734.1
Liquid
26998
FETUB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGLLLPLAPCILVLCCGAMSPPQLALNPSALLSRGCNDSDVLAVAGFALRDINKDRKDGYVLRLNRVNDAQEYRRGGLGSLFYLTLDVLETDCHVLRKKAWQDCGMRIFFESVYGQCKAIFYMNNPSRVLYLAAYNCTLRPVSKKKIYMTCPDCPSSIPTDSSNHQVLEAATESLAKYKNENTSKQYSLFKVTRASSQWVVGPSYFVEYLIKESPCTKSQASSCSLQSSDSVPVGLCKGSLTRTHWEKFVSVTCDFFESQAPATGSENSAVNQKPTNLPKVEESQQKNTPPTDSPSKAGPRGSVQYLPDLDDKNSQEKGPQEAFPVHLDLTTNPQGETLDISFLFLEPMEEKLVVLPFPKEKARTAECPGPAQNASPLVLPP
RUO
FETUB
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
16G2|Gugu|IRL685
FETUB
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit