Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FEZ1 Full-length ORF (NP_005094.1, 1 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name FEZ1 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 71.5kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-944-403
View Documents
Abnova™
H00009638P01L
25 ug
Each for $709.50
Add to cart
 
89-944-402
View Documents
Abnova™
H00009638P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

This gene is an ortholog of the C. elegans unc-76 gene, which is necessary for normal axonal bundling and elongation within axon bundles. Expression of this gene in C. elegans unc-76 mutants can restore to the mutants partial locomotion and axonal fasciculation, suggesting that it also functions in axonal outgrowth. The N-terminal half of the gene product is highly acidic. Alternatively spliced transcript variants encoding different isoforms of this protein have been described. [provided by RefSeq]

Sequence: MEAPLVSLDEEFEDLRPSCSEDPEEKPQCFYGSSPHHLEDPSLSELENFSSEIISFKSMEDLVNEFDEKLNVCFRNYNAKTENLAPVKNQLQIQEEEETLQDEEVWDALTDNYIPSLSEDWRDPNIEALNGNCSDTEIHEKEEEEFNEKSENDSGINEEPLLTADQVIEEIEEMMQNSPDPEEEEEVLEEEDGGETSSQADSVLLQEMQALTQTFNNNWSYEGLRHMSGSELTELLDQVEGAIRDFSEELVQQLARRDELEFEKEVKNSFITVLIEVQNKQKEQRELMKKRRKEKGLSLQSSRIEKGNQMPLKRFSMEGISNILQSGIRQTFGSSGTDKQYLNTVIPYEKKASPPSVEDLQMLTNILFAMKEDNEKVPTLLTDYILKVLCPT
Specifications

Specifications

FEZ1 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
71.5kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit