Learn More
Raybiotech IncSupplier Diversity Partner Human FGF-20 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHFGF201
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.3 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes human FGF-20. Other species not yet determined.
Catalog No. 50-194-9258