Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-6 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF238
This item is not returnable.
View return policy
Description
FGF-6 Polyclonal specifically detects FGF-6 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
FGF-6 | |
Polyclonal | |
Biotin | |
P10767 | |
FGF6 | |
E. coli-derived recombinant human FGF-6 (R&D Systems, Catalog # 238-F6) Gly41-Ile208 Accession # P10767 | |
50 μg | |
Primary | |
Detects human FGF-6 in ELISAs and Western blots. In sandwich immunoassays, less than 5% cross-reactivity with recombinant human (rh) FGF-4 is observed and less than 0.6% cross-reactivity with rhFGF acidic, rhFGF basic, rhFGF-5, rhFGF-7, rhFGF-9, rhFGF-10, rhFGF-17, and rhFGF-19 is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
FGF6, FGF-6, fibroblast growth factor 6, HBGF-6, Heparin secretory-transforming protein 2, Heparin-binding growth factor 6, HST2, HST-2, HSTF2, HSTF-2 | |
Goat | |
Affinity Purified | |
RUO | |
2251 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction