Learn More
Abnova™ Human FGF1 (P05230, 140 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3964
Catalog No. 89-944-428
Add to cart
Description
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis. Multiple alternatively spliced variants encoding different isoforms have been described. [provided by RefSeq]
Specifications
Specifications
P05230
Lyophilized
2246
FGF1 (Human) Recombinant Protein
≥98% by SDS-PAGE and HPLC
50 ug
RUO
AFGF|ECGF|ECGF-beta|ECGFA|ECGFB|FGF-alpha|FGFA|GLIO703|HBGF1
FGF1
The ED50 was determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors is < 10ng/mL, corresponding to a specific activity of > 1 x 105 units/mg.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
15.8kDa
Escherichia coli expression system
SDS-PAGE Result
Store at -20°C on dry atmosphere. After reconstitution with 10 mM Tris, store at 4°C for 1 week or -20°C for longer. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
FGF1
Escherichia coli expression system
Escherichia coli
None