Learn More
Abnova™ Human FGF17 (O60258) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5984
Catalog No. 89-944-449
Add to cart
Description
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was shown to be prominently expressed in the cerebellum and cortex. The mouse homolog of this gene was localized to specific sites in the midline structures of the forebrain, the midbrain-hindbrain junction, developing skeleton and developing arteries, which suggests a role in central nervous system, bone and vascular development. This gene was referred to as FGF-13 in reference 2, however, its amino acid sequence and chromosomal localization are identical to FGF17. [provided by RefSeq]
Sequence: MTQGENHPSPNFNQYVRDQGAMTDQLSRRQIREYQLYSRTSGKHVQVTGRRISATAEDGNKFAKLIVETDTFGSRVRIKGAESEKYICMNKRGKLIGKPSGKSKDCVFTEIVLENNYTAFQNARHEGWFMAFTRQGRPRQASRSRQNQREAHFIKRLYQGQLPFPNHAEKQKQFEFVGSAPTRRTKRTRRPQPLT