Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FGF5 Full-length ORF (NP_149134.1, 1 a.a. - 123 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002250P01L

 View more versions of this product

Catalog No. 89-944-486


Add to cart

Description

Description

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was identified as an oncogene, which confers transforming potential when transfected into mammalian cells. Targeted disruption of the homolog of this gene in mouse resulted in the phenotype of abnormally long hair, which suggested a function as an inhibitor of hair elongation. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MSLSFLLLLFFSHLILSAWAHGEKRLAPKGQPGPAATDRNPRGSSSRQSSSSAMSSSSASSSPAASLGSQGSGLEQSSFQWSPSGRRTGSLYCRVGIGFHLQIYPDGKVNGSHEANMLSQVHR
Specifications

Specifications

NP_149134.1
Liquid
2250
FGF5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSLSFLLLLFFSHLILSAWAHGEKRLAPKGQPGPAATDRNPRGSSSRQSSSSAMSSSSASSSPAASLGSQGSGLEQSSFQWSPSGRRTGSLYCRVGIGFHLQIYPDGKVNGSHEANMLSQVHR
RUO
FGF5
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HBGF-5|Smag-82
FGF5
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit