Learn More
Abnova™ Human FGF7 (P21781, 32 a.a. - 194 a.a.) Partial Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P3645
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein is a potent epithelial cell-specific growth factor, whose mitogenic activity is predominantly exhibited in keratinocytes but not in fibroblasts and endothelial cells. Studies of mouse and rat homologs of this gene implicated roles in morphogenesis of epithelium, reepithelialization of wounds, hair development and early lung organogenesis. [provided by RefSeq]
Sequence: CNDMTPEQMATNVNCSSPERHTRSYDYMEGGDIRVRRLFCRTQWYLRIDKRGKVKGTQEMKNNYNIMEIRTVAVGIVAIKGVESEFYLAMNKEGKLYAKKECNEDCNFKELILENHYNTYASAKWTHNGGEMFVALNQKGIPVRGKKTKKEQKTAHFLPMAITSpecifications
P21781 | |
Lyophilized | |
19kDa | |
Escherichia coli expression system | |
10 ug | |
Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg (1 EU/μg) | |
FGF7 | |
The ED50 was determined by the dose-dependent proliferation of BAF3 cells was found to be less than 10ng/mL. | |
Recombinant | |
Escherichia coli expression system | |
>90% by SDS-PAGE and HPLC |
Functional Study, SDS-PAGE | |
2252 | |
FGF7 (Human) Recombinant Protein | |
Ion exchange column and HPLC reverse phase column | |
CNDMTPEQMATNVNCSSPERHTRSYDYMEGGDIRVRRLFCRTQWYLRIDKRGKVKGTQEMKNNYNIMEIRTVAVGIVAIKGVESEFYLAMNKEGKLYAKKECNEDCNFKELILENHYNTYASAKWTHNGGEMFVALNQKGIPVRGKKTKKEQKTAHFLPMAIT | |
RUO | |
HBGF-7/KGF | |
FGF7 | |
E. coli | |
None | |
Lyophilized |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.