Learn More
Abnova™ Human FGF9 (P31371, 1 a.a. - 208 a.a.) Full-length Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P3608
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis. [provided by RefSeq]
Sequence: MAPLGEVGNYFGVQDAVPFGNVPVLPVDSPVLLSDHLGQSEAGGLPRGPAVTDLDHLKGILRRRQLYCRTGFHLEIFPNGTIQGTRKDHSRFGILEFISIAVGLVSIRGVDSGLYLGMNEKGELYGSEKLTQECVFREQFEENWYNTYSSNLYKHVDTGRRYYVALNKDGTPREGTRTKRHQKFTHFLPRPVDPDKVPELYKDILSQSSpecifications
P31371 | |
Lyophilized | |
23kDa | |
Escherichia coli expression system | |
20 ug | |
Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg (1 EU/μg) | |
FGF9 | |
The ED50 was determined by the dose-dependent proliferationof BaF3 cells expressing FGF receptors and was found to be in the range of 0.6ng/mL. | |
Recombinant | |
Escherichia coli expression system | |
>90% by SDS-PAGE and HPLC |
Functional Study, SDS-PAGE | |
2254 | |
FGF9 (Human) Recombinant Protein | |
Ion exchange column and HPLC reverse phase column | |
MAPLGEVGNYFGVQDAVPFGNVPVLPVDSPVLLSDHLGQSEAGGLPRGPAVTDLDHLKGILRRRQLYCRTGFHLEIFPNGTIQGTRKDHSRFGILEFISIAVGLVSIRGVDSGLYLGMNEKGELYGSEKLTQECVFREQFEENWYNTYSSNLYKHVDTGRRYYVALNKDGTPREGTRTKRHQKFTHFLPRPVDPDKVPELYKDILSQS | |
RUO | |
GAF/HBFG-9/MGC119914/MGC119915 | |
FGF9 | |
E. coli | |
None | |
Lyophilized |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.