Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FGFR4 (NP_002002.3, 460 a.a. - 802 a.a.) V550E mutant Partial Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5758

Catalog No. 89-944-539


Add to cart

Description

Description

The protein encoded by this gene is a member of the fibroblast growth factor receptor family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein would consist of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. The genomic organization of this gene, compared to members 1-3, encompasses 18 exons rather than 19 or 20. Although alternative splicing has been observed, there is no evidence that the C-terminal half of the IgIII domain of this protein varies between three alternate forms, as indicated for members 1-3. This particular family member preferentially binds acidic fibroblast growth factor and, although its specific function is unknown, it is overexpressed in gynecological tumor samples, suggesting a role in breast and ovarian tumorigenesis. [provided by RefSeq]

Specifications

Specifications

NP_002002.3
Liquid
2264
FGFR4 (V550E) (Human) Recombinant Protein
Glutathione sepharose chromatography
SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD334|JTK2|MGC20292|TKF
FGFR4
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : CSKtide. ATP: 100 µM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
66kDa
Baculovirus infected insect cell (Sf21) expression system
86% by SDS-PAGE/CBB staining
5 ug
RUO
FGFR4
Baculovirus infected insect cell (Sf21) expression system
Insect
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit