Learn More
Abnova™ Human FGR (NM_005248.1, 1 a.a. - 529 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4694
Catalog No. 89-944-557
Add to cart
Description
Description
This gene is a member of the Src family of protein tyrosine kinases (PTKs). The encoded protein contains N-terminal sites for myristylation and palmitylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. The protein localizes to plasma membrane ruffles, and functions as a negative regulator of cell migration and adhesion triggered by the beta-2 integrin signal transduction pathway. Infection with Epstein-Barr virus results in the overexpression of this gene. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Specifications
Specifications
NM_005248.1
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
93.934kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
FLJ43153|MGC75096|SRC2|c-fgr|c-src2|p55c-fgr|p58c-fgr
FGR
16pmol/μg x min
Yes
0.055 ug/uL
Liquid
2268
FGR (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
FGR
Insect cell (Sf9) expression system
Insect
GST-His