Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FIBP Full-length ORF (AAH14388, 1 a.a. - 357 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009158P01S

 View more versions of this product

Catalog No. 89-944-578


Add to cart

Description

Description

Acidic fibroblast growth factor is mitogenic for a variety of different cell types and acts by stimulating mitogenesis or inducing morphological changes and differentiation. The FIBP protein is an intracellular protein that binds selectively to acidic fibroblast growth factor (aFGF). It is postulated that FIBP may be involved in the mitogenic action of aFGF. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MTSELDIFVGNTTLIDEDVYRLWLDGYSVTDAVALRVRSGILEQTGATAAVLQSDTMDHYRTFHMLERLLHAPPKLLHQLIFQIPPSRQALLIERYYAFDEAFVREVLGKKLSKGTKKDLDDISTKTGITLKSCRRQFDNFKRVFKVVEEMRGSLVDNIQQHFLLSDRLARDYAAIVFFANNRFETGKKKLQYLSFGDFAFCAELMIQNWTLGAVDSQMDDMDMDLDKEFLQDLKELKVLVADKDLLDLHKSLVCTALRGKLGVFSEMEANFKNLSRGLVNVAAKLTHNKDVGDLFVDLVEKFVEPCRSDHWPLSDVRFFLNQYSASVHSLDGFRHQALWDRYMGTLRGCLLRLYHD
Specifications

Specifications

AAH14388
Liquid
9158
FIBP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTSELDIFVGNTTLIDEDVYRLWLDGYSVTDAVALRVRSGILEQTGATAAVLQSDTMDHYRTFHMLERLLHAPPKLLHQLIFQIPPSRQALLIERYYAFDEAFVREVLGKKLSKGTKKDLDDISTKTGITLKSCRRQFDNFKRVFKVVEEMRGSLVDNIQQHFLLSDRLARDYAAIVFFANNRFETGKKKLQYLSFGDFAFCAELMIQNWTLGAVDSQMDDMDMDLDKEFLQDLKELKVLVADKDLLDLHKSLVCTALRGKLGVFSEMEANFKNLSRGLVNVAAKLTHNKDVGDLFVDLVEKFVEPCRSDHWPLSDVRFFLNQYSASVHSLDGFRHQALWDRYMGTLRGCLLRLYHD
RUO
FIBP
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.01kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FGFIBP|FIBP-1
FIBP
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit