Learn More
Novus Biologicals™ Human Fibrinogen beta chain ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP267955
Catalog No. NBP267955
Add to Cart
Description
Description
The Human Fibrinopeptide B ELISA Kit (Colorimetric) measures human Fibrinopeptide B in serum, plasma and other biological fluids.
Specifications
Specifications
1.56 - 100 ng/mL
This kit recognizes Human FG(beta) in samples. No significant cross-reactivity or interference between Human FG(beta) and analogues was observed.
Biological Fluids, Plasma, Serum
ELISA
FGB
1 Kit
Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78 p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78 p, MGC104327, MGC120405
0.94 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Human Fibrinogen beta chain in samples. No significant cross-reactivity or interference between Human Fibrinogen beta chain and analogues was observed.
2244
Human
Storage of components varies. See protocol for specific instructions.
Fibrinogen beta chain
Recommendations
Recommendations