Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FIG4 Full-length ORF (NP_055660.1, 1 a.a. - 907 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name FIG4 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 130kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-949-673
View Documents
Abnova™
H00009896P01L
25 ug
Each for $709.50
Add to cart
 
89-949-672
View Documents
Abnova™
H00009896P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

The protein encoded by this gene belongs to the SAC domain-containing protein gene family. The SAC domain, approximately 400 amino acids in length and consisting of seven conserved motifs, has been shown to possess phosphoinositide phosphatase activity. The yeast homolog, Sac1p, is involved in the regulation of various phosphoinositides, and affects diverse cellular functions such as actin cytoskeleton organization, Golgi function, and maintenance of vacuole morphology. Membrane-bound phosphoinositides function as signaling molecules and play a key role in vesicle trafficking in eukaryotic cells. Mutations in this gene have been associated with Charcot-Marie-Tooth disease, type 4J. [provided by RefSeq]

Sequence: MPTAAAPIISSVQKLVLYETRARYFLVGSNNAETKYRVLKIDRTEPKDLVIIDDRHVYTQQEVRELLGRLDLGNRTKMGQKGSSGLFRAVSAFGVVGFVRFLEGYYIVLITKRRKMADIGGHAIYKVEDTNMIYIPNDSVRVTHPDEARYLRIFQNVDLSSNFYFSYSYDLSHSLQYNLTVLRMPLEMLKSEMTQNRQESFDIFEDEGLITQGGSGVFGICSEPYMKYVWNGELLDIIKSTVHRDWLLYIIHGFCGQSKLLIYGRPVYVTLIARRSSKFAGTRFLKRGANCEGDVANEVETEQILCDASVMSFTAGSYSSYVQVRGSVPLYWSQDISTMMPKPPITLDQADPFAHVAALHFDQMFQRFGSPIIILNLVKEREKRKHERILSEELVAAVTYLNQFLPPEHTIVYIPWDMAKYTKSKLCNVLDRLNVIAESVVKKTGFFVNRPDSYCSILRPDEKWNELGGCVIPTGRLQTGILRTNCVDCLDRTNTAQFMVGKCALAYQLYSLGLIDKPNLQFDTDAVRLFEELYEDHGDTLSLQYGGSQLVHRVKTYRKIAPWTQHSKDIMQTLSRYYSNAFSDADRQDSINLFLGVFHPTEGKPHLWELPTDFYLHHKNTMRLLPTRRSYTYWWTPEVIKHLPLPYDEVICAVNLKKLIVKKFHKYEEEIDIHNEFFRPYELSSFDDTFCLAMTSSARDFMPKTVGIDPSPFTVRKPDETGKSVLGNKSNREEAVLQRKTAASAPPPPSEEAVSSSSEDDSGTDREEEGSVSQRSTPVKMTDAGDSAKVTENVVQPMKELYGINLSDGLSEEDFSIYSRFVQLGQSQHKQDKNSQQPCSRCSDGVIKLTPISAFSQDNIYEVQPPRVDRKSTEIFQAHIQASQGIMQPLGKEDSSMYREYIRNRYL
Specifications

Specifications

FIG4 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
130kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit