Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FIGLA Partial ORF (NP_001004311.1, 110 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00344018Q01L

 View more versions of this product

Catalog No. 89-944-583


Add to cart

Description

Description

This gene encodes a protein that functions in postnatal oocyte-specific gene expression. The protein is a basic helix-loop-helix transcription factor that regulates multiple oocyte-specific genes, including genes involved in folliculogenesis and those that encode the zona pellucida. Mutations in this gene cause premature ovarian failure type 6. [provided by RefSeq]

Sequence: ATEYIQVLSDLLEGAKDSKKQDPDEQSYSNNTSESHTSSARQLSRNITQHISCAFGLKNEEEGPWADGGSGEPAHACRHSVMSTTEIISPTRSLDRFPEVELLSHRLPQV
Specifications

Specifications

NP_001004311.1
Liquid
344018
FIGLA (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
POF6|bHLHc8
FIGLA
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ATEYIQVLSDLLEGAKDSKKQDPDEQSYSNNTSESHTSSARQLSRNITQHISCAFGLKNEEEGPWADGGSGEPAHACRHSVMSTTEIISPTRSLDRFPEVELLSHRLPQV
RUO
FIGLA
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit