Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FLJ10774 Partial ORF (NP_078938, 2 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Specifications

Name Human FLJ10774 Partial ORF (NP_078938, 2 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal
Recombinant Yes
For Use With (Application) AP, Array, ELISA, WB-Re
Molecular Weight 36.41kDa
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-944-654
View Documents
Abnova™
H00055226Q01L
25μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
89-944-653
View Documents
Abnova™
H00055226Q01S
10μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Sequence: HRKKVDNRIRILIENGVAERQRSLFVVVGDRGKDQVVILHHMLSKATVKARPSVLWCYKKELGFSSHRKKRMRQLQKKIKNGTLNIKQDDPFELFIA
Specifications

Specifications

Human FLJ10774 Partial ORF (NP_078938, 2 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal
AP, Array, ELISA, WB-Re
Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Yes
36.41kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit