Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human FNBP1L Partial ORF (NP_060207.2, 175 a.a. - 239 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00054874Q01L
Description
The protein encoded by this gene binds to both CDC42 and N-WASP. This protein promotes CDC42-induced actin polymerization by activating the N-WASP-WIP complex and, therefore, is involved in a pathway that links cell surface signals to the actin cytoskeleton. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: LNLRTHMADENKNEYAAQLQNFNGEQHKHFYVVIPQIYKQLQEMDERRTIKLSECYRGFADSERKSpecifications
NP_060207.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.89kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LNLRTHMADENKNEYAAQLQNFNGEQHKHFYVVIPQIYKQLQEMDERRTIKLSECYRGFADSERK | |
RUO | |
FNBP1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54874 | |
FNBP1L (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C1orf39/TOCA1 | |
FNBP1L | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction