Learn More
Abnova™ Human FOXA3 Full-length ORF (NP_004488.2, 1 a.a. - 350 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00003171P01S
Description
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. The crystal structure of a similar protein in rat has been resolved. [provided by RefSeq]
Sequence: MLGSVKMEAHDLAEWSYYPEAGEVYSPVTPVPTMAPLNSYMTLNPLSSPYPPGGLPASPLPSGPLAPPAPAAPLGPTFPGLGVSGGSSSSGYGAPGPGLVHGKEMPKGYRRPLAHAKPPYSYISLITMAIQQAPGKMLTLSEIYQWIMDLFPYYRENQQRWQNSIRHSLSFNDCFVKVARSPDKPGKGSYWALHPSSGNMFENGCYLRRQKRFKLEEKVKKGGSGAATTTRNGTGSAASTTTPAATVTSPPQPPPPAPEPEAQGGEDVGALDCGSPASSTPYFTGLELPGELKLDAPYNFNHPFSINNLMSEQTPAPPKLDVGFGGYGAEGGEPGVYYQGLYSRSLLNASSpecifications
NP_004488.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKHH3/HNF3G/MGC10179/TCF3G | |
FOXA3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3171 | |
FOXA3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLGSVKMEAHDLAEWSYYPEAGEVYSPVTPVPTMAPLNSYMTLNPLSSPYPPGGLPASPLPSGPLAPPAPAAPLGPTFPGLGVSGGSSSSGYGAPGPGLVHGKEMPKGYRRPLAHAKPPYSYISLITMAIQQAPGKMLTLSEIYQWIMDLFPYYRENQQRWQNSIRHSLSFNDCFVKVARSPDKPGKGSYWALHPSSGNMFENGCYLRRQKRFKLEEKVKKGGSGAATTTRNGTGSAASTTTPAATVTSPPQPPPPAPEPEAQGGEDVGALDCGSPASSTPYFTGLELPGELKLDAPYNFNHPFSINNLMSEQTPAPPKLDVGFGGYGAEGGEPGVYYQGLYSRSLLNAS | |
RUO | |
FOXA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.