Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FRAT1 Partial ORF (NP_005470.2, 181 a.a. - 279 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010023Q01L

 View more versions of this product

Catalog No. 89-945-126


Add to cart

Description

Description

The protein encoded by this gene belongs to the GSK-3-binding protein family. The protein inhibits GSK-3-mediated phosphorylation of beta-catenin and positively regulates the Wnt signaling pathway. It may function in tumor progression and in lymphomagenesis. [provided by RefSeq]

Sequence: RLQQRRGSQPETRTGDDDPHRLLQQLVLSGNLIKEAVRRLHSRRLQLRAKLPQRPLLGPLSAPVHEPPSPRSPRAACSDPGASGRAQLRTGDGVLVPGS
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue
RLQQRRGSQPETRTGDDDPHRLLQQLVLSGNLIKEAVRRLHSRRLQLRAKLPQRPLLGPLSAPVHEPPSPRSPRAACSDPGASGRAQLRTGDGVLVPGS
RUO
FRAT1
Wheat Germ (in vitro)
GST
NP_005470.2
Liquid
10023
FRAT1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ97193
FRAT1
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit