Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FTO Full-length ORF (AAH01284.1, 1 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00079068P01S

 View more versions of this product

Catalog No. 89-945-193


Add to cart

Description

Description

The exact function of this gene is not know. Studies in mice suggest that it may be involved in nucleic acid demethylation, and that its mRNA level is regulated by feeding and fasting. Genomewide association studies of type 2 diabetes indicate this gene as a diabetes susceptibility locus. Mutation in this gene has been associated with growth retardation, developmental delay, coarse facies, and early death. [provided by RefSeq]

Sequence: MGHPRAIQPSVFFSPYDVHFLLYPIRCPYLKIGRFHIKLKGLHFLFSFLFFFFETQSHSVTRLECSGTISAHCNLCLPGSSNSPASASQVAGTTGTCHHAQLIFVFLAEMGFHHIGQDGLDLNLVIHPPRSPKALGLQA
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
41.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1752/MGC5149
FTO
Yes
AAH01284.1
Liquid
79068
FTO (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGHPRAIQPSVFFSPYDVHFLLYPIRCPYLKIGRFHIKLKGLHFLFSFLFFFFETQSHSVTRLECSGTISAHCNLCLPGSSNSPASASQVAGTTGTCHHAQLIFVFLAEMGFHHIGQDGLDLNLVIHPPRSPKALGLQA
RUO
FTO
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit