Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FXC1 Full-length ORF (AAH11014, 1 a.a. - 103 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026515P01S

 View more versions of this product

Catalog No. 89-945-261


Add to cart

Description

Description

FXC1, or TIMM10B, belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane.[supplied by OMIM]

Sequence: MERQQQQQQQLRNLRDFLLVYNRMTELCFQRCVPSLHHRALDAEEEACLHSCAGKLIHSNHRLMAAYVQLMPALVQRRIADYEAASTVPSVAAEQPGVSPSGS
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.07kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TIM10B/TIMM10B/Tim9b
FXC1
Yes
AAH11014
Liquid
26515
FXC1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MERQQQQQQQLRNLRDFLLVYNRMTELCFQRCVPSLHHRALDAEEEACLHSCAGKLIHSNHRLMAAYVQLMPALVQRRIADYEAASTVPSVAAEQPGVSPSGS
RUO
FXC1
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit