Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FZD1 (aa 73-253), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

CHO cells Recombinant Protein

$102.80 - $738.50

Specifications

Recombinant Yes
Name Human FZD1 (aa 73-253)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87822 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87822
10 µg
Each for $102.80
Add to cart
 
PIRP87823 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87823
25 µg
Each for $212.00
Add to cart
 
PIRP87824 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87824
100 µg
Each for $738.50
Add to cart
 
Description

Description

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD1 protein contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, 7 transmembrane domains, and a C-terminal PDZ domain-binding motif. The FZD1 transcript is expressed in various tissues.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative; pH 7.2
CHO cells
-20°C
Q9UP38
Fzd1
>95%
Human FZD1 (aa 73-253)
200 µg/mL
Liquid
AW227548; Chfz-1; Dfrizzled-1; DKFZp564G072; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 1; FLJ95923; frizzled; frizzled 1; frizzled 1, seven transmembrane spanning receptor; frizzled class receptor 1; frizzled family receptor 1; frizzled homolog 1; frizzled homolog 1 (Drosophila); frizzled, Drosophila, homolog of, 1; frizzled-1; Fz receptor 1; Fz1; FZ-1; Fzd1; FzE1; hFz1; mFz1; rFz1; Wnt receptor; Xfrizzled-1
RUO
Human
8321
46.6 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit