Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human Gage12B Full-Length Orf (Adr82812.1, 1 A.A.-117 A.A.) Recombinant Protein With Gst Tag At N-Terminal.

Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.

$360.00 - $548.50

Specifications

Format Liquid
Regulatory Status RUO
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
Accession Number ADR82812.1
Purification Method Glutathione Sepharose 4 Fast Flow
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-032-109 View Documents Abnova™
H00729428P01S
10 ug Each for $360.00
Add to cart
 
89-032-110 View Documents Abnova™
H00729428P01L
25 ug Each for $548.50
Add to cart
 
Description

Description

  • Good Folding
  • Excellent amino acids labeling

Immunogen Sequence: MSWRGRSTYYWPRPRRYVQPPEMIGPMRPEQFSDEVEPATPEEGEPATQCQDPAAAQEGEDEGASAGQGPKPEAHSQEQGHPQTGCECEDGPDGQEMDPPNPEEVKTPEEGEKQSQC

Specifications

Specifications

Liquid
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
Glutathione Sepharose 4 Fast Flow
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RUO
ADR82812.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit