Learn More
FUJIFILM IRVINE SCIENTIFIC INC Human Galectin-1 , Recombinant Protein, Shenandoah Biotechnology, 50 ug vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 1003050UG
Encompass_Preferred
Galectin-1 belongs to the lectin family of carbohydrate binding proteins and binds glycans as both a monomer and a homodimer. Galectin-1 is produced in peripheral lymphoid organs and inflammatory sites. Galectin-1 plays important roles in acute and chronic inflammatory processes, cell growth, cell proliferation, and induces apoptosis of activated T cells. Galectin-1 also modulates cytokine secretion and inhibits pro-inflammatory cytokine production.
Catalog No. 50-990-741
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.