Learn More
Novus Biologicals™ Human Galectin-3 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191275
Catalog No. NBP191275
Add to cart
Description
Description
The Human Galectin-3 ELISA Kit (Colorimetric) measures human Galectin-3 in cell culture supernatant, serum, plasma (edta).
Specifications
Specifications
0.39 - 25.0ng/mL
3958
ELISA Kit
0.12ng/mL
Human (Hu)
Galectin-3
LGALS3
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Galectin-3, Biotin-Conjugate anti-human Galectin-3 polyclonal antibody, Streptavidin-HRP, Human Galectin-3 Standard lyophilized, 50ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA)
Storage of components varies. See protocol for specific instructions.
Apoptosis, Cancer, Cellular Markers, Tumor Suppressors
Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-bi