Learn More
Raybiotech IncSupplier Diversity Partner Human Galectin-8 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHGALECTIN81
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.16 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Galectin-8 (LGALS8)
Catalog No. 50-194-9290