Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GALNT10 Partial ORF (NP_938080, 503 a.a. - 602 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055568Q01L

 View more versions of this product

Catalog No. 89-945-455


Add to cart

Description

Description

This gene belongs to the polypeptide N-acetylgalactosaminyltransferase (pp-GalNAc-T) gene family. Polypeptide GalNAc transferases initiate the synthesis of mucin-type oligosaccharides by transferring GalNAc from UDP-GalNAc to the hydroxyl group of either a serine or threonine residue on the polypeptide acceptor. Following expression in insect cells, recombinant GalNAc transferase 10 showed significant GalNAcT activity toward mucin-derived peptides, and it utilized both nonglycosylated and glycosylated peptide substrates. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: FTFTWREDIRPGDPQHTKKFCFDAISHTSPVTLYDCHSMKGNQLWKYRKDKTLYHPVSGSCMDCSESDHRIFMNTCNPSSLTQQWLFEHTNSTVLEKFNR
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FTFTWREDIRPGDPQHTKKFCFDAISHTSPVTLYDCHSMKGNQLWKYRKDKTLYHPVSGSCMDCSESDHRIFMNTCNPSSLTQQWLFEHTNSTVLEKFNR
RUO
GALNT10
Wheat Germ (in vitro)
GST
NP_938080
Liquid
55568
GALNT10 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp586H0623/FLJ00205/FLJ11715/GalNAcT10/pp-GalNAc-T10
GALNT10
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit