Learn More
Raybiotech IncSupplier Diversity Partner Human GATA-1/Eryf1 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHGATA11
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 16 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human GATA-1.
Catalog No. 50-194-9297