Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GDAP1L1 Full-length ORF (NP_076939.3, 1 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name GDAP1L1 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 68.4kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-945-685
View Documents
Abnova™
H00078997P01L
25 ug
Each for $709.50
Add to cart
 
89-945-684
View Documents
Abnova™
H00078997P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

The ganglioside GD3 synthase causes cell differentiation with neurite sprouting when transfected into the mouse neuroblastoma cell line Neuro2a. After differentiation, the expression of several genes is upregulated, including one that encodes a protein termed ganglioside-induced differentiation-associated protein 1 (Gdap1). A similar gene was found in humans, and mutations in the human gene are associated with Charcot-Marie-Tooth type 4A disease. The protein encoded by this gene is similar in sequence to the human GDAP1 protein. [provided by RefSeq]

Sequence: MATPNNLTPTNCSWWPISALESDAAKPAEAPDAPEAASPAHWPRESLVLYHWTQSFSSQKVRLVIAEKGLVCEERDVSLPQSEHKEPWFMRLNLGEEVPVIIHRDNIISDYDQIIDYVERTFTGEHVVALMPEVGSLQHARVLQYRELLDALPMDAYTHGCILHPELTTDSMIPKYATAEIRRHLANATTDLMKLDHEEEPQLSEPYLSKQKKLMAKILEHDDVSYLKKILGELAMVLDQIEAELEKRKLENEGQKCELWLCGCAFTLADVLLGATLHRLKFLGLSKKYWEDGSRPNLQSFFERVQRRFAFRKVLGDIHTTLLSAVIPNAFRLVKRKPPSFFGASFLMGSLGGMGYFAYWYLKKKYI
Specifications

Specifications

GDAP1L1 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
68.4kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit