Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GDF-9 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8266V100UG
This item is not returnable.
View return policy
Description
GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Flow Cytometry.Specifications
GDF-9 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
Human embryonic kidney cell line derived recombinant human GDF-9 Met1-Arg454 Accession # O60383 | |
RUO | |
2661 | |
Human | |
IgG2b |
Flow Cytometry | |
917319 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O60383 | |
Mouse | |
100 μg | |
Primary | |
Detects human GDF-9 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction