Learn More
Raybiotech IncSupplier Diversity Partner Human GDF-9 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHGDF91
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 2.2 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human GDF-9.
Catalog No. 50-194-9307