Learn More
Pepro Tech Inc Animal-Free Recombinant Human GDF-5 (BMP-14/CDMP-1), 250ug DFS Item
Manufacturer:  Pepro Tech Inc AF12001250UG
Recombinant Human GDF-5 (BMP-14/CDMP-1) is a 27.0 kDa homodimeric disulfide-linked protein consisting of two 120 amino acid polypeptide chains.
Catalog No. 50-195-2634