Learn More
R&D Systems™ Human GDNF Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity
$5230.00
Specifications
Product Type Recombinant protein
Regulatory Status RUO
Recombinant Yes
Biological Activity 2 to 12ng/mL
Quantity 1mg
View More Specs
Products 2
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
21-2GD-01M View Documents R&D Systems™
212GD01M
Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein Each for $5,230.00
Add to cart
21-2GD-01MCF View Documents R&D Systems™
212GD01M/CF
Lyophilized from a 0.2μm filtered solution in PBS Each for $5,230.00
Add to cart
Description
Description
The Recombinant Human GDNF Protein is derived from NS0. The Recombinant Human GDNF Protein has been validated for the following applications: Bioactivity, Binding Activity.
Order Info
Ships at ambient temperature.
Specifications
Specifications
Recombinant protein
Yes
1mg
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<1.0 EU per 1μg of the protein by the LAL method
Mouse myeloma cell line, NS0-derived Arg109-Ile211
RUO
2 to 12ng/mL
Unlabeled
Human
11.6 kDa (monomer)
P39905