Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human GFR alpha-3/GDNF R alpha-3 Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Product Type Recombinant Protein
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol GFRA3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB5965210UG View Documents Novus Biologicals™
NBP25965210UG
10μg Each for $287.00
Add to cart
 
NB5965250UG View Documents Novus Biologicals™
NBP25965250UG
50μg Each for $1,025.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 32-374 of Human GFR alpha-3/GDNF R alpha-3 The Recombinant Human GFR alpha-3/GDNF R alpha-3 Protein is derived from Baculovirus. The Recombinant Human GFR alpha-3/GDNF R alpha-3 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Recombinant Protein
RUO
GFRA3
GFRA3 Recombinant Protein
In PBS (pH 7.4) containing 10% glycerol
Baculovirus
Unlabeled
2676
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
GDNF family receptor alpha 3, GDNF family receptor alpha-3, GDNF R alpha-3, GDNF receptor alpha-3, GDNFR-alpha-3, GFR alpha3
>85% by SDS-PAGE
Human
PAGE
65.5 kDa

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit