Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GluR6/GRIK2 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9610S100UG
This item is not returnable.
View return policy
Description
KLHL24 Monoclonal antibody specifically detects KLHL24 in Human samples. It is validated for Flow CytometrySpecifications
KLHL24 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
DRE1, EAA4, GLR6, GluK2, GLUK6, Kainate Receptor Interacting Protein For GluR6, Kainate Receptor-Interacting Protein For GluR6, Kelch-Like 24, Kelch-Like 24 (Drosophila), Kelch-Like Family Member 24, Kelch-Like Protein 24, KLHL24, KRIP6, MRT6, Protein DRE1 | |
Chinese Hamster Ovary cell line CHO-derived human GluR6/GRIK2 protein, Thr32-Asn549, Accession # Q13002 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
818235 | |
Flow Cytometry | |
Q13002 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
54800 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2a |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction