Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Glypican 3 Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Product Type Recombinant Protein
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol GPC3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB59651250U View Documents Novus Biologicals™
NBP259651250UG
250μg Each for $1,025.00
Add to cart
 
NB5965150UG View Documents Novus Biologicals™
NBP25965150UG
50μg Each for $287.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 25-559 of Human Glypican 3 The Recombinant Human Glypican 3 Protein is derived from Baculovirus. The Recombinant Human Glypican 3 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Recombinant Protein
RUO
GPC3
GPC3 Recombinant Protein
In PBS (pH 7.4) containing 10% glycerol
Baculovirus
Unlabeled
2719
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
DGSX, Glypican 3, glypican proteoglycan 3, glypican-3, GPC3, GTR2-2
>90% by SDS-PAGE
Human
PAGE
61.8 kDa

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit