Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human GMF-beta Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$89.50 - $5770.00
Specifications
Accession Number | P60983 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 2764 |
Molecular Weight (g/mol) | 16.5 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
45037100UG
|
Gibco™
45037100UG |
100 μg |
Each for $1,154.00
|
|
|||||
450-37-10UG
|
Gibco™
4503710UG |
10 μg |
Each for $217.00
|
|
|||||
450-37-1MG
|
Gibco™
450371MG |
1 mg |
Each for $5,770.00
|
|
|||||
45037250UG
|
Gibco™
45037250UG |
250 μg |
Each for $2,020.00
|
|
|||||
450-37-2UG
|
Gibco™
450372UG |
2 μg |
Each for $89.50
|
|
|||||
45037500UG
|
Gibco™
45037500UG |
500 μg |
Each for $3,460.00
|
|
|||||
Description
This protein, which is phosphorylated following phorbol ester stimulation, is important for the nervous system. It causes brain cell differentiation, stimulates neural regeneration and inhibits tumor cell proliferation. Overexpression of GMF in astrocytes has been shown to enhance brain-derived neurotrohic factor (BDNF) production. GMF expression is increased by exercise, and the protein is crucial for exercise-induction of BDNF. Through BDNF production, GMF appears to play a role in neuroprotection. In thymoma, T-cell development is maintained by GMF; being produced by the tumor cells.Specifications
P60983 | |
protein with no preservative | |
16.5 | |
E. coli | |
RUO | |
3110001H22Rik; 3110001O16Rik; AI851627; C79176; D14Ertd630e; DNA for thyroid hormone receptor binding site (276 bp); glia maturation factor beta; glia maturation factor beta-like protein; glia maturation factor, beta; GMF; Gmfb; GMF-beta | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
2764 | |
Human GMF-beta | |
-20°C | |
<1 EU/μg | |
Gmfb | |
Unconjugated | |
SESLVVCDVA EDLVEKLRKF RFRKETNNAA IIMKIDKDKR LVVLDEELEG ISPDELKDEL PERQPRFIVY SYKYQHDDGR VSYPLCFIFS SPVGCKPEQQ MMYAGSKNKL VQTAELTKVF EIRNTEDLTE EWLREKLGFF H | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title